3-Bromo-1,8-naphthalic anhydride 97%

Supplier: Thermo Scientific Chemicals
Warning

L17683-03
CAAAAL17683-03EA 173.8 CAD
CAAAAL17683-03 CAAAAL17683-MD
3-Bromo-1,8-naphthalic anhydride 97%
3-Bromo-1,8-naphthalic anhydride

Purity: 97% CAS No.: 24050-49-5 Molecular Formula: C12H5BrO3 Formula Weight: 277.08 Melting Point: 244-246° Beilstein Reference: 180001

Formula: C₁₂H₅BrO₃
Melting Pt: 244…246 °C
Storage Temperature: Ambient
MDL Number: MFCD00228162
CAS Number: 24050-49-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR