Boron trifluoride tetrahydrofuran complex BF₃ 45.5%

Supplier: Thermo Scientific Chemicals
Danger

L17593-22 L17593-36
CAAAAL17593-22EA 56.88 CAD
CAAAAL17593-22 CAAAAL17593-36
Boron trifluoride tetrahydrofuran complex BF₃ 45.5%
Boron trifluoride tetrahydrofuran complex

CAS No.: 462-34-0 Molecular Formula: C4H8BF3O Formula Weight: 139.91 Flash Point: 92°(197°F) Melting Point: 8-9° Boiling Point: 180° UN No.: 2922 MDL No.: MFCD00040372

Formula: C₄H₈BF₃O
Boiling Pt: 180 °C
Melting Pt: 8…9 °C
Density: 1.268
Flash Pt: 92 °C (197 °F)
Storage Temperature: Ambient
MDL Number: MFCD00040372
CAS Number: 462-34-0
EINECS: 207-325-9
UN: 3265
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR