N-(5-Bromopentyl)phthalimide 97%

Supplier: Thermo Scientific Chemicals

L17543-06 L17543-14 L17543-22
CAAAAL17543-06EA 72.68 CAD
CAAAAL17543-06 CAAAAL17543-14 CAAAAL17543-22
N-(5-Bromopentyl)phthalimide 97%
N-(5-Bromopentyl)phthalimide

Purity: 97% CAS No.: 954-81-4 Molecular Formula: C13H14BrNO2 Formula Weight: 296.17 Odor: Odorless Melting Point: 58-62° Beilstein Reference: 177015 MDL No.: MFCD00060522

Formula: C₁₃H₁₄BrNO₂
MW: 296.16 g/mol
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00060522
CAS Number: 954-81-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR