5-(Trifluoromethoxy)salicylaldehyde 98+%

Supplier: Thermo Scientific Chemicals
Warning

L17136-03
CAAAAL17136-03EA 254.38 CAD
CAAAAL17136-03 CAAAAL17136-MD
5-(Trifluoromethoxy)salicylaldehyde 98+%
5-(Trifluoromethoxy)salicylaldehyde

Purity: 98+% CAS No.: 93249-62-8 Molecular Formula: C8H5F3O3 Formula Weight: 206.12 Flash Point: 110°(230°F) Melting Point: 31-33° Boiling Point: 82°/60mm Beilstein Reference: 7023825 MDL No.: MFCD00075249

Formula: C₈H₅F₃O₃
Boiling Pt: 82 °C/60 mm
Melting Pt: 31…33 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075249
CAS Number: 93249-62-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR