1H,1H,2H-Perfluoro-1-hexene 97%

Supplier: Thermo Scientific Chemicals
Danger

L16834-06 L16834-14 L16834-22
CAAAAL16834-06EA 57.99 CAD
CAAAAL16834-06 CAAAAL16834-14 CAAAAL16834-22
1H,1H,2H-Perfluoro-1-hexene 97%
1H,1H,2H-Perfluoro-1-hexene

Purity: 97% CAS No.: 19430-93-4 Molecular Formula: C6H3F9 Formula Weight: 246.08 Flash Point: None Refractive Index: 1.2890 Boiling Point: 59-60° Beilstein Reference: 1819716 MDL No.: MFCD00042338

Formula: C₆H₃F₉
Boiling Pt: 59…60 °C
Density: 1.457
Storage Temperature: Ambient
MDL Number: MFCD00042338
CAS Number: 19430-93-4
EINECS: 243-053-7
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR