2,4-Dichloro-6-methylbenzylamine 98+%

Supplier: Thermo Scientific Chemicals
Danger

L16108-03
CAAAAL16108-03EA 183.28 CAD
CAAAAL16108-03 CAAAAL16108-06 CAAAAL16108-MD
2,4-Dichloro-6-methylbenzylamine 98+%
2,4-Dichloro-6-methylbenzylamine

Purity: 98+% CAS No.: 150517-76-3 Molecular Formula: C8H9Cl2N Formula Weight: 190.07 Melting Point: 52-56° Beilstein Reference: 8615227 UN No.: 3259 MDL No.: MFCD00052913

Formula: C₈H₉Cl₂N
Melting Pt: 52…56 °C
Storage Temperature: Ambient
MDL Number: MFCD00052913
CAS Number: 150517-76-3
UN: 3259
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR