4-Amino-2-chlorobenzonitrile 98+%

Supplier: Thermo Scientific Chemicals
Warning

L15924-06 L15924-14 L15924-22
CAAAAL15924-06EA 49.14 CAD
CAAAAL15924-06 CAAAAL15924-14 CAAAAL15924-22
4-Amino-2-chlorobenzonitrile 98+%
4-Amino-2-chlorobenzonitrile

Purity: 98+% CAS No.: 20925-27-3 Molecular Formula: C7H5ClN2 Formula Weight: 152.58 Melting Point: 116-119° Beilstein Reference: 2936425 UN No.: 3439 MDL No.: MFCD00035926

Formula: C₇H₅ClN₂
Boiling Pt: 301 °C (1013 hPa)
Melting Pt: 116…119 °C
Storage Temperature: Ambient
MDL Number: MFCD00035926
CAS Number: 20925-27-3
EINECS: 244-114-0
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR