Ethyl cyanoformate 99%

Supplier: Thermo Scientific Chemicals
Danger

L15777-06 L15777-14 L15777-22
CAAAAL15777-06EA 64.94 CAD
CAAAAL15777-06 CAAAAL15777-14 CAAAAL15777-22
Ethyl cyanoformate 99%
Ethyl cyanoformate

Purity: 99% CAS No.: 623-49-4 Molecular Formula: NCCO2CH2CH3 Formula Weight: 99.09 Flash Point: 24°(75°F) Refractive Index: 1.3820 Boiling Point: 115-116° Beilstein Reference: 635955 UN No.: 3275 MDL No.: MFCD00001836

Formula: C₄H₅NO₂
Boiling Pt: 115…116 °C
Density: 1.003
Flash Pt: 24 °C(75 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001836
CAS Number: 623-49-4
EINECS: 210-797-9
UN: 2929
ADR: "6.1",II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR