Methylenediphenyl-4,4'-diisocyanate 98% stabilized

Supplier: Thermo Scientific Chemicals
Danger

L15740-14 L15740-22 L15740-36
CAAAAL15740-14EA 47.08 CAD
CAAAAL15740-14 CAAAAL15740-22 CAAAAL15740-36
Methylenediphenyl-4,4'-diisocyanate 98% stabilized
Methylenediphenyl-4,4'-diisocyanate

Purity: 98% CAS No.: 101-68-8 Molecular Formula: C15H10N2O2 Formula Weight: 250.26 Storage Temperature: -30°C to -10°C Flash Point: 196°(385°F) Odor: Weak, characteristic Melting Point: 41-44° Boiling Point: 194°/5mm Beilstein Reference: 797662 UN No.: 2811 MDL No.: MFCD00036131

Formula: C₁₅H₁₀N₂O₂
Boiling Pt: 194 °C/5 mm
Melting Pt: 41…44 °C
Density: 1.239
Flash Pt: 196 °C(385 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00036131
CAS Number: 101-68-8
EINECS: 202-966-0
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR