Phosphorus pentoxide - methanesulfonic acid (∼7.5% P₂O₅) (w/w)

Supplier: Thermo Scientific Chemicals
Danger

L15607-AE L15607-AP
CAAAAL15607-AEEA 89.11 CAD
CAAAAL15607-AE CAAAAL15607-AP
Phosphorus pentoxide - methanesulfonic acid (∼7.5% P₂O₅) (w/w)
Phosphorus pentoxide - methanesulfonic acid

Fieser: 5,535 7,291 8,400 11,428 20,309


CAS No.: 39394-84-8 Molecular Formula: O5P2 Formula Weight: 141.95 Flash Point: 110°(230°F) Refractive Index: 1.4350 UN No.: 3265 MDL No.: MFCD00070545

Density: 1.515
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00070545
CAS Number: 39394-84-8
UN: 2922
ADR: "8",III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR