(R)-(+)-2-Methyl-CBS-oxazaborolidine 1 M in toluene

Supplier: Thermo Scientific Chemicals
Danger

L14582-AA L14582-AC
CAAAAL14582-AAEA 138.25 CAD
CAAAAL14582-AA CAAAAL14582-AC CAAAAL14582-XF
(R)-(+)-2-Methyl-CBS-oxazaborolidine 1 M in toluene
(R)-(+)-2-Methyl-CBS-oxazaborolidine

Fieser: 14,239 16,253 17,242


CAS No.: 112022-83-0 Molecular Formula: C18H20BNO Formula Weight: 277.17 Flash Point: 4°(39°F) UN No.: 1993 MDL No.: MFCD00078440

Formula: C₁₈H₂₀BNO
Density: 0.925
Flash Pt: 4 °C (39 °F)
Storage Temperature: Ambient
MDL Number: MFCD00078440
CAS Number: 112022-83-0
UN: 1294
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR