Methyl-3-bromopropionate 97%

Supplier: Thermo Scientific Chemicals
Warning

L14127-14 L14127-22
CAAAAL14127-14EA 107.28 CAD
CAAAAL14127-14 CAAAAL14127-22
Methyl-3-bromopropionate 97%
Methyl 3-bromopropionate

Purity: 97% CAS No.: 3395-91-3 Molecular Formula: Br(CH2)2CO2CH3 Formula Weight: 167.01 Flash Point: 75°(167°F) Refractive Index: 1.4580 Boiling Point: 58-60°/12mm Beilstein Reference: 1742949 MDL No.: MFCD00000250

Formula: C₄H₇BrO₂
Boiling Pt: 58…60 °C/12 mm
Density: 1.530
Flash Pt: 75 °C(167 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000250
CAS Number: 3395-91-3
EINECS: 222-247-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR