1-Boc-piperazine 99%

Supplier: Thermo Scientific Chemicals
Warning

L13363-03 L13363-06 L13363-14
CAAAAL13363-03EA 55.62 CAD
CAAAAL13363-03 CAAAAL13363-06 CAAAAL13363-14
1-Boc-piperazine 99%
1-Boc-piperazine

Purity: 99% CAS No.: 57260-71-6 Molecular Formula: C9H18N2O2 Formula Weight: 186.26 Melting Point: 44-48° Beilstein Reference: 879985

Formula: C₉H₁₈N₂O₂
Melting Pt: 44…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00075265
CAS Number: 57260-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR