Acridine orange (hemizinc salt)

Supplier: Thermo Scientific Chemicals
Warning

L13159-06 L13159-14
CAAAAL13159-06EA 51.03 CAD
CAAAAL13159-06 CAAAAL13159-14
Acridine orange (hemizinc salt)
Acridine orange (hemizinc salt)

Fluorescent stain for proteins. An RNA polymerase inhibitor.


CAS No.: 10127-02-3 Molecular Formula: C17H19N3.HCl.ZnCl2 Formula Weight: 438.10 Beilstein Reference: 3734978 MDL No.: MFCD00081043

Formula: C₁₇H₂₀ClN₃·HCl·½ZnCl₂
Storage Temperature: Ambient
MDL Number: MFCD00081043
CAS Number: 10127-02-3
EINECS: 233-353-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR