Sodium carbonate 98%

Supplier: Thermo Scientific Chemicals
Warning

L13098-0E L13098-0I L13098-36
CAAAAL13098-0EEA 132.88 CAD
CAAAAL13098-0E CAAAAL13098-0I CAAAAL13098-36
Sodium carbonate 98%
Sodium carbonate

Purity: 98% CAS No.: 497-19-8 Molecular Formula: Na2CO3 Formula Weight: 105.99 Refractive Index: 1.535 Odor: Odorless pH: 11,5 Beilstein Reference: 4154566 Merck Reference: 14,8596 MDL No.: MFCD00003494

Formula: Na₂CO₃
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 854 °C
Density: 2.53
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,8596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR