Ethyl-4-(trifluoromethyl)benzoate 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 4-(trifluoromethyl)benzoate

L11139-03
CAAAAL11139-03EA 40.13 CAD
CAAAAL11139-03
Ethyl-4-(trifluoromethyl)benzoate 99%
Ethyl 4-(trifluoromethyl)benzoate

Purity: 99% CAS No.: 583-02-8 Molecular Formula: C10H9F3O2 Formula Weight: 218.18 Refractive Index: 1.4470 Boiling Point: 78-80°/5mm MDL No.: MFCD00013563

Formula: C₁₀H₉F₃O₂
MW: 218.18 g/mol
Boiling Pt: 78…80 °C/5 mm
Storage Temperature: Ambient
MDL Number: MFCD00013563
CAS Number: 583-02-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR