4-(2-Pyridylazo)resorcinol 99%

Supplier: Thermo Scientific Chemicals
Warning

L09667-03 L09667-06
CAAAAL09667-03EA 47.56 CAD
CAAAAL09667-03 CAAAAL09667-06
4-(2-Pyridylazo)resorcinol 99%
4-(2-Pyridylazo)resorcinol

Purity: 99% CAS No.: 1141-59-9 Molecular Formula: C11H9N3O2 Formula Weight: 215.21 Melting Point: ca 187° dec. Beilstein Reference: 184665

Formula: C₁₁H₉N₃O₂
Melting Pt: ca 187 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00044237
CAS Number: 1141-59-9
EINECS: 214-528-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR