3-(Trimethylsilyl)-2-propyn-1-ol 98+%

Supplier: Thermo Scientific Chemicals
Warning

L09649-03 L09649-06 L09649-14
CAAAAL09649-03EA 62.73 CAD
CAAAAL09649-03 CAAAAL09649-06 CAAAAL09649-14
3-(Trimethylsilyl)-2-propyn-1-ol 98+%
3-(Trimethylsilyl)-2-propyn-1-ol

Purity: 98+% CAS No.: 5272-36-6 Molecular Formula: C6H12OSi Formula Weight: 128.25 Storage Temperature: 2°C to 8°C Flash Point: 66°(150°F) Refractive Index: 1.4510 Boiling Point: 170-172° Beilstein Reference: 1902505 UN No.: 2810 MDL No.: MFCD00002913

Formula: C₆H₁₂OSi
Boiling Pt: 170…172 °C
Density: 0.865
Flash Pt: 66 °C(150 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00002913
CAS Number: 5272-36-6
EINECS: 226-094-5
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR