2,3,4,5-Tetramethyl-2-cyclopentenone cis- and trans-mixture 95%

Supplier: Thermo Scientific Chemicals
Warning

L09617-06 L09617-14
CAAAAL09617-06EA 116.76 CAD
CAAAAL09617-06 CAAAAL09617-14
2,3,4,5-Tetramethyl-2-cyclopentenone cis- and trans-mixture 95%
2,3,4,5-Tetramethyl-2-cyclopentenone

Purity: 95% CAS No.: 54458-61-6 Molecular Formula: C9H14O Formula Weight: 138.21 Flash Point: 73°(163°F) Refractive Index: 1.4760 Boiling Point: 99-100°/30mm Beilstein Reference: 2324088 MDL No.: MFCD00010248

Formula: C₉H₁₄O
Boiling Pt: 99…100 °C/30 mm
Density: 0.917
Flash Pt: 73 °C (163 °F)
Storage Temperature: Ambient
MDL Number: MFCD00010248
CAS Number: 54458-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR