D(-)-Lysine monohydrochloride 98%

Supplier: Thermo Scientific Chemicals

L07710-03 L07710-06
CAAAAL07710-03EA 60.67 CAD
CAAAAL07710-03 CAAAAL07710-06
D(-)-Lysine monohydrochloride 98%
D(-)-Lysine monohydrochloride

Purity: 98% CAS No.: 7274-88-6 Molecular Formula: C6H14N2O2.HCl Formula Weight: 182.65 Melting Point: ca 260° dec. Beilstein Reference: 4356907

Formula: C₆H₁₅ClN₂O₂
Melting Pt: ∼ 260 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012920
CAS Number: 7274-88-6
EINECS: 230-691-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR