3-(Trifluoromethyl)thiophenol 97%

Supplier: Thermo Scientific Chemicals
Warning

L07191-03 L07191-06
CAAAAL07191-03EA 75.37 CAD
CAAAAL07191-03 CAAAAL07191-06
3-(Trifluoromethyl)thiophenol 97%
3-(Trifluoromethyl)thiophenol

Purity: 97% CAS No.: 937-00-8 Molecular Formula: C7H5F3S Formula Weight: 178.18 Refractive Index: 1.4900 Odor: Stench Boiling Point: 71-72°/20mm Beilstein Reference: 1943114 UN No.: 2810 MDL No.: MFCD00041142

Formula: C₇H₅F₃S
Boiling Pt: 71…72 °C/20 mm
Storage Temperature: Ambient
MDL Number: MFCD00041142
CAS Number: 937-00-8
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR