Oxypurinol ≥98%

Supplier: Thermo Scientific Chemicals
Danger

L07160-03 L07160-06 L07160-MD
CAAAAL07160-03EA 139.67 CAD
CAAAAL07160-03 CAAAAL07160-06 CAAAAL07160-MD
Oxypurinol ≥98%
Oxypurinol

An inhibitor of xanthine oxidase


Purity: 98+% CAS No.: 2465-59-0 Molecular Formula: C5H4N4O2 Formula Weight: 152.11 Odor: Odorless Melting Point: 300° Beilstein Reference: 139956 MDL No.: MFCD00056934

Formula: C₅H₄N₄O₂
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00047205
CAS Number: 2465-59-0
EINECS: 219-570-9
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR