1-Heptanethiol 98%

Supplier: Thermo Scientific Chemicals
Warning

L07079-14 L07079-22
CAAAAL07079-14EA 83.9 CAD
CAAAAL07079-14 CAAAAL07079-22
1-Heptanethiol 98%
1-Heptanethiol

Purity: 98% CAS No.: 1639-09-4 Molecular Formula: CH3(CH2)6SH Formula Weight: 132.27 Flash Point: 46°(114°F) Refractive Index: 1.4515 Melting Point: -43° Boiling Point: 173-175° Beilstein Reference: 1731687 UN No.: 3336 MDL No.: MFCD00004911

Formula: C₇H₁₆S
Boiling Pt: 173…175 °C
Melting Pt: –43 °C
Density: 0.854
Flash Pt: 46 °C (114 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004911
CAS Number: 1639-09-4
EINECS: 216-678-8
UN: 3336
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR