4-Ethoxycinnamic acid predominantly trans ≥98%

Supplier: Thermo Scientific Chemicals
Warning

L06324-06 L06324-14
CAAAAL06324-06EA 126.4 CAD
CAAAAL06324-06 CAAAAL06324-14
4-Ethoxycinnamic acid predominantly trans ≥98%
4-Ethoxycinnamic acid

Purity: 98+% CAS No.: 2373-79-7 Molecular Formula: C11H12O3 Formula Weight: 192.22 Melting Point: 195-199° Beilstein Reference: 3198068 MDL No.: MFCD00016848

Formula: C₁₁H₁₂O₃
Melting Pt: 195…199 °C
Storage Temperature: Ambient
MDL Number: MFCD11040581
CAS Number: 2373-79-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR