(±)-2-Hexanol 99%

Supplier: Thermo Scientific Chemicals

L05993-14 L05993-22
CAAAAL05993-14EA 96.22 CAD
CAAAAL05993-14 CAAAAL05993-22
(±)-2-Hexanol 99%
(±)-2-Hexanol

Purity: 99% CAS No.: 626-93-7 Molecular Formula: C6H14O Formula Weight: 102.18 Flash Point: 45°(113°F) Refractive Index: 1.4140 Odor: Characteristic Melting Point: -23° Boiling Point: 137-139° Beilstein Reference: 1718996 UN No.: 2282 MDL No.: MFCD00004585

Formula: C₆H₁₄O
Storage Temperature: Ambient
MDL Number: MFCD00004585
CAS Number: 626-93-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9db7ec887376d519b077785cdead15c1aed20d58&vid=5c0c76fb523ec80ddb5134157d5d19ce7a63dcc3&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR