Pentyl-4-hydroxybenzoate 98%

Supplier: Thermo Scientific Chemicals

L05563-06 L05563-14
CAAAAL05563-06EA 70.15 CAD
CAAAAL05563-06 CAAAAL05563-14
Pentyl-4-hydroxybenzoate 98%
Pentyl 4-hydroxybenzoate

Purity: 98% CAS No.: 6521-29-5 Molecular Formula: C12H16O3 Formula Weight: 208.26 Melting Point: 36-40° Boiling Point: 197-200°/10mm MDL No.: MFCD00016479

Formula: C₁₂H₁₆O₃
Boiling Pt: 197…200 °C (10 mmHg)
Melting Pt: 36…40 °C
Storage Temperature: Ambient
MDL Number: MFCD00016479
CAS Number: 6521-29-5
EINECS: 229-408-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR