4-Ethoxybenzhydrazide 98+%

Supplier: Thermo Scientific Chemicals
Warning

L05433-06
CAAAAL05433-06EA 114.08 CAD
CAAAAL05433-06
4-Ethoxybenzhydrazide 98+%
4-Ethoxybenzhydrazide

Purity: 98+% CAS No.: 58586-81-5 Molecular Formula: C9H12N2O2 Formula Weight: 180.21 Melting Point: 124-127° Beilstein Reference: 2642900

Formula: C₉H₁₂N₂O₂
Melting Pt: 124…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00017074
CAS Number: 58586-81-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR