6-Methoxy-2(3H)-benzoxazolone ≥98%

Supplier: Thermo Scientific Chemicals

Synonyms: Coixol, 6-Methoxybenzo[d]oxazol-2(3H)-one

L05349-03 L05349-MD
CAAAAL05349-03EA 229.1 CAD
CAAAAL05349-03 CAAAAL05349-MD
6-Methoxy-2(3H)-benzoxazolone ≥98%
6-Methoxy-2(3H)-benzoxazolone

Purity: 98+% CAS No.: 532-91-2 Molecular Formula: C8H7NO3 Formula Weight: 165.15 Melting Point: 152-156° Beilstein Reference: 141041 MDL No.: MFCD00051726

Formula: C₈H₇NO₃
MW: 165.15 g/mol
Melting Pt: 152…156 °C
Storage Temperature: Ambient
MDL Number: MFCD00051726
CAS Number: 532-91-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR