2-Methylbenzylamine 98%

Supplier: Thermo Scientific Chemicals
Danger

L05243-06 L05243-14
CAAAAL05243-06EA 141.41 CAD
CAAAAL05243-06 CAAAAL05243-14
2-Methylbenzylamine 98%
2-Methylbenzylamine

Purity: 98% CAS No.: 89-93-0 Molecular Formula: C8H11N Formula Weight: 121.18 Flash Point: 83°(181°F) Refractive Index: 1.5440 Boiling Point: 198-200° Beilstein Reference: 1099406 UN No.: 2735 MDL No.: MFCD00008112

Formula: C₈H₁₁N
Boiling Pt: 198…200 °C
Melting Pt: < 0 °C
Density: 0.979
Flash Pt: 83 °C (181 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008112
CAS Number: 89-93-0
EINECS: 201-952-1
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR