Bisacrylamide 97%

Supplier: Thermo Scientific Chemicals
Warning

L04694-22 L04694-36
CAAAAL04694-22EA 60.04 CAD
CAAAAL04694-22 CAAAAL04694-36
Bisacrylamide 97%
Bisacrylamide

Purity: 97% CAS No.: 110-26-9 Molecular Formula: C7H10N2O2 Formula Weight: 154.17 Storage Temperature: 2°C to 8°C Melting Point: 300° Beilstein Reference: 1706297 MDL No.: MFCD00008625

Formula: (H₂C=CHCONH)₂CH₂
Boiling Pt: 125 °C (3.33 hPa)
Melting Pt: >300 °C
Density: 1.235
Flash Pt: 138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008625
CAS Number: 110-26-9
EINECS: 203-750-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1b72b0c3cd5bb916c65f9e2337c5c3b05ea1c91&vid=368f6bf0bc8e0b35ffc7a57d85f5dd210ff0b409&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR