1-Pentanethiol 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: n-Pentanethiol, 1-Mercaptopentane, n-Pentyl mercaptan

L04330-14
CAAAAL04330-14EA 105.86 CAD
CAAAAL04330-14
1-Pentanethiol 97%
1-Pentanethiol

Purity: 97% CAS No.: 110-66-7 Molecular Formula: CH3(CH2)4SH Formula Weight: 104.22 Flash Point: 18°(64°F) Refractive Index: 1.4460 Odor: Stench Melting Point: -76° Boiling Point: 125-126° Beilstein Reference: 1730979 UN No.: 1111 MDL No.: MFCD00004907

Formula: C₅H₁₂S
MW: 104.22 g/mol
Boiling Pt: 127
Melting Pt: -76
Density: 0.841
Flash Pt: 18 °C (64 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004907
CAS Number: 110-66-7
EINECS: 203-789-1
UN: 1111
ADR: 3 II
Merck Index: 12,00653

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR