4-Bromobenzoyl chloride 98+%

Supplier: Thermo Scientific Chemicals
Danger

L03794-09 L03794-18
CAAAAL03794-09EA 64.78 CAD
CAAAAL03794-09 CAAAAL03794-18
4-Bromobenzoyl chloride 98+%
4-Bromobenzoyl chloride

Purity: 98+% CAS No.: 586-75-4 Molecular Formula: C7H4BrClO Formula Weight: 219.47 Flash Point: 110°(230°F) Melting Point: 38-42° Boiling Point: 246° Beilstein Reference: 636641 UN No.: 3261 MDL No.: MFCD00000683

Formula: C₇H₄BrClO
Boiling Pt: 246 °C
Melting Pt: 38…42 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000683
CAS Number: 586-75-4
EINECS: 209-580-1
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR