Ethyl nicotinate 99%

Supplier: Thermo Scientific Chemicals
Warning

L03215-22 L03215-36
CAAAAL03215-22EA 69.05 CAD
CAAAAL03215-22 CAAAAL03215-36
Ethyl nicotinate 99%
Ethyl nicotinate

Purity: 99% CAS No.: 614-18-6 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Flash Point: 93°(199°F) Refractive Index: 1.5030 Odor: Characteristic Melting Point: 8-9° Boiling Point: 223-224° Beilstein Reference: 122937 MDL No.: MFCD00006389

Formula: C₈H₉NO₂
Boiling Pt: 223…224 °C
Melting Pt: 8…9 °C
Density: 1,107
Flash Pt: 93 °C(199 °F)
Storage Temperature: Ambient
MDL Number: MFCD00006389
CAS Number: 614-18-6
EINECS: 210-370-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR