2-Hydroxyethyl methyl sulfone 97%

Supplier: Thermo Scientific Chemicals

L03159-06
CAAAAL03159-06EA 89.59 CAD
CAAAAL03159-06
2-Hydroxyethyl methyl sulfone 97%
2-Hydroxyethyl methyl sulfone

Purity: 97% CAS No.: 15205-66-0 Molecular Formula: C3H8O3S Formula Weight: 124.16 Flash Point: 110°(230°F) Refractive Index: 1.4790 Melting Point: 27-30° Boiling Point: 148-149°/1mm Beilstein Reference: 1746915 MDL No.: MFCD00014747

Formula: C₃H₈O₃S
Boiling Pt: 148…149 °C (1 mmHg)
Melting Pt: 27…30 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00014747
CAS Number: 15205-66-0
EINECS: 239-261-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR