Tritylium hexafluorophosphate 98%

Supplier: Thermo Scientific Chemicals
Danger

L03140-09 L03140-18
CAAAAL03140-09EA 222.78 CAD
CAAAAL03140-09 CAAAAL03140-18
Tritylium hexafluorophosphate 98%
Tritylium hexafluorophosphate

Purity: 98% CAS No.: 437-17-2 Molecular Formula: C19H15F6P Formula Weight: 388.29 Storage Temperature: -30°C to -10°C Melting Point: ca 160° dec. Beilstein Reference: 4344297 UN No.: 3261 MDL No.: MFCD00013121

Formula: C₁₉H₁₅F₆P
Melting Pt: ∼ 160 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00013121
CAS Number: 437-17-2
EINECS: 207-112-0
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR