4-Bromophenylboronic acid 97+%

Supplier: Thermo Scientific Chemicals
Warning

L01565-03 L01565-06 L01565-14
CAAAAL01565-03EA 39.03 CAD
CAAAAL01565-03 CAAAAL01565-06 CAAAAL01565-14
4-Bromophenylboronic acid 97+%
4-Bromophenylboronic acid

Purity: 97+% CAS No.: 5467-74-3 Molecular Formula: C6H6BBrO2 Formula Weight: 200.83 Melting Point: 282-286° Beilstein Reference: 2936347

Formula: C₆H₆BBrO₂
Melting Pt: 282…286 °C
Density: 1,67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002104
CAS Number: 5467-74-3
EINECS: 226-779-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR