(1-Naphthoxy)acetic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

L01036-06 L01036-14
CAAAAL01036-06EA 67.62 CAD
CAAAAL01036-06 CAAAAL01036-14
(1-Naphthoxy)acetic acid 98+%
(1-Naphthoxy)acetic acid

Purity: 98+% CAS No.: 2976-75-2 Molecular Formula: C12H10O3 Formula Weight: 202.21 Melting Point: 193-197° Beilstein Reference: 1912165

Formula: C₁₂H₁₀O₃
Melting Pt: 193…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00003927
CAS Number: 2976-75-2
EINECS: 221-023-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR