2-Bromobenzhydrazide 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Bromobenzene-1-carbohydrazide, 2-Bromobenzohydrazide, 2-Bromobenzoic acid hydrazide, 2-Bromobenzoic hydrazide

L00937-06 L00937-14
CAAAAL00937-06EA 66.04 CAD
CAAAAL00937-06 CAAAAL00937-14
2-Bromobenzhydrazide 98+%
2-Bromobenzhydrazide

Purity: 98+% CAS No.: 29418-67-5 Molecular Formula: C7H7BrN2O Formula Weight: 215.05 Melting Point: 152-154° Beilstein Reference: 1941021

Formula: C₇H₇BrN₂O
Melting Pt: 152…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00051577
CAS Number: 29418-67-5
EINECS: 249-614-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR