Ethyl indole-2-carboxylate 98+%

Supplier: Thermo Scientific Chemicals

L00797-06 L00797-14
CAAAAL00797-06EA 81.69 CAD
CAAAAL00797-06 CAAAAL00797-14
Ethyl indole-2-carboxylate 98+%
Ethyl indole-2-carboxylate

Purity: 98+% CAS No.: 3770-50-1 Molecular Formula: C11H11NO2 Formula Weight: 189.21 Melting Point: 122-125° Beilstein Reference: 146395

Formula: C₁₁H₁₁NO₂
Melting Pt: 122…125 °C
Storage Temperature: Ambient
MDL Number: MFCD00005609
CAS Number: 3770-50-1
EINECS: 223-206-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR