Bromopyruvic acid 97%

Supplier: Thermo Scientific Chemicals
Danger

L00720-04 L00720-09 L00720-18
CAAAAL00720-04EA 62.57 CAD
CAAAAL00720-04 CAAAAL00720-09 CAAAAL00720-18
Bromopyruvic acid 97%
Bromopyruvic acid

Purity: 97% CAS No.: 1113-59-3 Molecular Formula: C3H3BrO3 Formula Weight: 166.96 Storage Temperature: 2°C to 8°C Melting Point: 75-80° Beilstein Reference: 1746786 UN No.: 3261 MDL No.: MFCD00002587

Formula: BrH₂CC(O)COOH
Boiling Pt: 249 °C (1013 hPa)
Melting Pt: 75…80 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002587
CAS Number: 1113-59-3
EINECS: 214-206-5
UN: 3261
ADR: 8,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR