Ethyl diethoxyacetate 97+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diethoxyacetic ethyl ester

L00390-14 L00390-22
CAAAAL00390-14EA 116.45 CAD
CAAAAL00390-14 CAAAAL00390-22 CAAAAL00390-36
Ethyl diethoxyacetate 97+%
Ethyl diethoxyacetate

Purity: 97+% CAS No.: 6065-82-3 Molecular Formula: C8H16O4 Formula Weight: 176.21 Flash Point: 85°(185°F) Refractive Index: 1.4100 Boiling Point: 198-200° Beilstein Reference: 878579 MDL No.: MFCD00009231

Formula: C₈H₁₆O₄
MW: 176.21 g/mol
Boiling Pt: 198…200 °C
Density: 0.988
Flash Pt: 85 °C (185 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009231
CAS Number: 6065-82-3
EINECS: 227-999-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR