Di-tert-butyl hydrazodicarboxylate ≥98%

Supplier: Thermo Scientific Chemicals
Warning

L00280-06 L00280-14
CAAAAL00280-06EA 103.33 CAD
CAAAAL00280-06 CAAAAL00280-14
Di-tert-butyl hydrazodicarboxylate ≥98%
Di-tert-butyl hydrazodicarboxylate

Purity: 98+% CAS No.: 16466-61-8 Molecular Formula: C10H20N2O4 Formula Weight: 232.28 Melting Point: 123-126° Beilstein Reference: 1794023 MDL No.: MFCD00015000

Formula: C₁₀H₂₀N₂O₄
Melting Pt: 123…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00015000
CAS Number: 16466-61-8
EINECS: 240-512-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR