Isopropyl glycolate 96%

Supplier: Thermo Scientific Chemicals
Warning

H50334-03 H50334-06
CAAAAH50334-03EA 164.32 CAD
CAAAAH50334-03 CAAAAH50334-06
Isopropyl glycolate 96%
Isopropyl glycolate

Purity: 96% CAS No.: 623-61-0 Molecular Formula: HOCH2CO2CH(CH3)2 Formula Weight: 118.13 MDL No.: MFCD00046747

Formula: HOCH₂CO₂CH(CH₃)₂
MW: 118.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00046747
CAS Number: 623-61-0
EINECS: 210-804-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR