Thieno[3,2-b]pyridin-7-ol 96%

Supplier: Thermo Scientific Chemicals
Warning

H50172-03 H50172-06
CAAAAH50172-03EA 83.9 CAD
CAAAAH50172-03 CAAAAH50172-06
Thieno[3,2-b]pyridin-7-ol 96%
Thieno[3,2-b]pyridin-7-ol

Purity: 96% CAS No.: 107818-20-2 Molecular Formula: C7H5NOS Formula Weight: 151.19 MDL No.: MFCD01863636

Formula: C₇H₅NOS
Storage Temperature: Ambient
MDL Number: MFCD01863636
CAS Number: 107818-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR