(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 98%

Supplier: Thermo Scientific Chemicals
Warning

B25251-06 B25251-14
CAAAAB25251-06EA 136.51 CAD
CAAAAB25251-06 CAAAAB25251-14
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP) 98%
(Benzotriazol-1-yloxy)tri(1-pyrrolidinyl)phosphonium hexafluorophosphate (PyBOP)

Purity: 98% CAS No.: 128625-52-5 Molecular Formula: C18H28F6N6OP2 Formula Weight: 520.40 Storage Temperature: 2°C to 8°C Melting Point: ca 155° dec. Beilstein Reference: 3584612 MDL No.: MFCD00077411

Formula: C₁₈H₂₈F₆N₆OP₂
Melting Pt: ∼ 155 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00077411
CAS Number: 128625-52-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR