6(5H)-Phenanthridinone 96%

Supplier: Thermo Scientific Chemicals

B25187-03 B25187-06
CAAAAB25187-03EA 64.94 CAD
CAAAAB25187-03 CAAAAB25187-06
6(5H)-Phenanthridinone 96%
6(5H)-Phenanthridinone

Purity: 96% CAS No.: 1015-89-0 Molecular Formula: C13H9NO Formula Weight: 195.22 Melting Point: 288-294° Beilstein Reference: 140641

Formula: C₁₃H₉NO
Melting Pt: 288…294 °C
Storage Temperature: Ambient
MDL Number: MFCD00004988
CAS Number: 1015-89-0
EINECS: 213-804-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR