Creatine (N-amidinosarcosine) monohydrate 99%

Supplier: Thermo Scientific Chemicals

Synonyms: Creatine Hydrate, 2-(1-Methylguanidino)acetic acid monohydrate, Creatine monohydrate, 2-(1-Methylguanidino)acetic acid hydrate, N-Amidinosarcosine monydrate

B25009-0E B25009-22 B25009-36
CAAAAB25009-0EEA 820.02 CAD
CAAAAB25009-0E CAAAAB25009-22 CAAAAB25009-36
Creatine (N-amidinosarcosine) monohydrate 99%
Creatine (N-Amidinosarcosine) monohydrate

Purity: 99% CAS No.: 6020-87-7 Molecular Formula: C4H9N3O2.H2O Formula Weight: 149.15 (131.13anhy) Odor: Weak pH: 6.9 Melting Point: ca 292° dec. Beilstein Reference: 907175 Merck Reference: 14,2568 MDL No.: MFCD00071582

Formula: C₄H₉N₃O₂·1H₂O
MW: 149.15 g/mol (131.13 anhy)
Melting Pt: ∼ 292 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00071582
CAS Number: 6020-87-7
EINECS: 200-306-6
Merck Index: 14,2568

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR