2,4-Dichlorobenzyl mercaptan 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (2,4-Dichlorophenyl)methanethiol, 2,4-Dichlorobenzenemethanethiol

B24999-06 B24999-14
CAAAAB24999-06EA 98.59 CAD
CAAAAB24999-06 CAAAAB24999-14
2,4-Dichlorobenzyl mercaptan 98+%
2,4-Dichlorobenzyl mercaptan

Purity: 98+% CAS No.: 59293-67-3 Molecular Formula: C7H6Cl2S Formula Weight: 193.10 Flash Point: 100°(212°F) Refractive Index: 1.6045 Boiling Point: 150-151°/12mm Beilstein Reference: 509602 UN No.: 2810 MDL No.: MFCD00039650

Formula: C₇H₆Cl₂S
MW: 193.10 g/mol
Boiling Pt: 150…151 °C (12 mmHg)
Density: 1.36
Flash Pt: >100 °C (212 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039650
CAS Number: 59293-67-3
EINECS: 261-687-2
UN: 2810
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR