5-Nitroisatin 98%

Supplier: Thermo Scientific Chemicals
Warning

B24962-06 B24962-14
CAAAAB24962-06EA 63.99 CAD
CAAAAB24962-06 CAAAAB24962-14 CAAAAB24962-22
5-Nitroisatin 98%
5-Nitroisatin

Purity: 98% CAS No.: 611-09-6 Molecular Formula: C8H4N2O4 Formula Weight: 192.13 Melting Point: ca 252° dec. Beilstein Reference: 180223

Formula: C₈H₄N₂O₄
Melting Pt: ∼ 252 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005720
CAS Number: 611-09-6
EINECS: 210-252-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR