2-Methylpyridin-3-ol 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Hydroxy-2-methylpyridine

B24944-03 B24944-06 B24944-14
CAAAAB24944-03EA 50.56 CAD
CAAAAB24944-03 CAAAAB24944-06 CAAAAB24944-14
2-Methylpyridin-3-ol 99%
2-Methylpyridin-3-ol

Purity: 99% CAS No.: 1121-25-1 Molecular Formula: C6H7NO Formula Weight: 109.13 Odor: Odorless Melting Point: 166-169° Beilstein Reference: 107937 MDL No.: MFCD00082538

Formula: C₆H₇NO
MW: 109.13 g/mol
Melting Pt: 166…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00082538
CAS Number: 1121-25-1
EINECS: 214-327-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR